Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Galactosidase Antibody (13R4), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP281034AF700
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Flow Cytometry.Specifications
beta Galactosidase | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rat | |
Protein A purified | |
RUO | |
13R4 recognizes wild-type E. coli beta-galactosidase. | |
Store at 4°C in the dark. | |
IgG1 λ |
Western Blot, Flow Cytometry | |
13R4 | |
Western Blot, Flow Cytometry | |
EC 3.2.1.23; b-gal | |
beta Galactosidase. | |
0.1 mL | |
Primary | |
E. coli | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction