Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Galactosidase Antibody (13R4), Alexa Fluor™ 750, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP252702AF750

Catalog No. NP22702A70


Only null left
Add to Cart

Description

Description

beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

beta Galactosidase
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
Mouse
0.1 mL
13R4 recognizes wild-type E. coli beta-galactosidase.
Store at 4°C in the dark.
Western Blot, Flow Cytometry
13R4
Western Blot, Flow Cytometry
EC 3.2.1.23; b-gal
Beta-galactosidase.
Primary
E. coli
IgG1 λ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.