Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Galactosidase Antibody (13R4), HRP, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP281033H
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot.Specifications
beta Galactosidase | |
Monoclonal | |
HRP | |
PBS | |
Armenian Hamster | |
Protein A purified | |
RUO | |
13R4 recognizes wild-type E. coli beta-galactosidase. | |
Store at 4°C in the dark. | |
IgG λ |
Western Blot | |
13R4 | |
Western Blot | |
EC 3.2.1.23; b-gal | |
beta Galactosidase. | |
0.1 mL | |
Primary | |
E. coli | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction