Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Galactosidase Antibody (BG-02) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500334 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500334 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500334 Supplier Novus Biologicals Supplier No. NB500334
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen beta Galactosidase
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone BG-02
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Accession No. Q56987
Gene Alias EC 3.2.1.23; b-gal
Host Species Mouse
Immunogen beta-Galactosidase purified from E. coli.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Beta Galactosidase Epitope Tags, Epitope Tags, Fusion Proteins
Primary or Secondary Primary
Test Specificity The antibody BG-02 recognizes a non-catalytic centre epitope of beta-galactosidase from E. coli.
Target Species E. coli
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.