Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Galactosidase Antibody (BG-02), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB359372 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359372 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359372 Supplier Novus Biologicals Supplier No. NB500334MFV500
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

beta Galactosidase Monoclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen beta Galactosidase
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone BG-02
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 3.2.1.23; b-gal
Host Species Mouse
Immunogen beta-Galactosidase purified from E. coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species E. coli
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.