Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Galactosidase Antibody (DC1 4C7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002619H
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
beta Galactosidase | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein A purified | |
RUO | |
E. coli | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
DC1 4C7 | |
Western Blot, ELISA, Immunohistochemistry | |
EC 3.2.1.23; b-gal | |
Beta-galactosidase EC.3.2.1.23 from E.coli | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction