Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ beta-Galactosidase Assay Stop Solution

Catalog No. 75706
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-706 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-706 Supplier Thermo Scientific™ Supplier No. 75706
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Thermo Scientific™ Mammalian β-Galactosidase Assay Stop Solution is a component of the Mammalian β-Galactosidase Assay Kit, which provides a simple colorimetric method for lysing cultured mammalian cells and measuring beta-Gal activity.

The Thermo Scientific™ Mammalian β-Galactosidase Assay Stop Solution is a component of the Mammalian β-Galactosidase Assay Kit, which provides a simple colorimetric method for lysing cultured mammalian cells and measuring beta-Gal activity.

The Assay Reagent and Kit comprise a simple and versatile assay system for measuring β-galactosidase activity. Mammalian cells grown on a plate may be lysed and tested for β-galactosidase activity by adding a single, all-in-one lysis-and-assay solution. Alternatively, the included Thermo Scientific™ M-PER Mammalian Protein Extraction Reagent may be used to prepare lysate from washed adherent or pelleted cells, and then a portion of the lysate used to quantitate the β-Gal activity. With M-PER Reagent, there is no need to scrape cells or to perform freeze-thaw cycles. The remaining lysates prepared with M-PER Reagent may be used for applications such as reporter assays (luciferase, β-galactosidase and chloramphenical acetyltransferase), protein kinase assays (PKA, PKC, and Tyrosine Kinase), immunoassays (Western blot, ELISA, RIA) and protein purification.

Features of the Thermo Scientific™ Mammalian β-Galactosidase Assay Kit (assay reagent and stop solution):

  • Choose stand-alone assay reagent or complete kit for cell lysis and detection
  • Colorimetric assay easily measured in a plate reader or spectrophotometer (405 nm)
  • Adaptable to 96-well microplates or tube assay formats
  • Perform homogeneous screening assays or measure activity secondarily after cell lysis
  • Also compatiible with prepared lysates from other species such as bacteria

Related Products

  • Mammalian beta-Galactosidase Assay Kit (Cat. No. 75707)
  • beta-Galactosidase Assay Reagent (Cat. No. 75705)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Detection Method NULL
For Use With (Application) Absorbance
For Use With (Equipment) Microplate Reader
Product Type Beta-Galactosidase Assay Stop Solution
Dye Type Enzyme Labeled
Format 96-well Plate
Product Line Pierce
Quantity 25 mL
Substrate Proprietary
Substrate Type Beta-Gal Substrate
Target Beta-Galactosidase
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.