Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Galactosidase Chimeric Recombinant Human Monoclonal Antibody (13R4)
GREENER_CHOICE

Catalog No. PIMA547784
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547784 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. PIMA547784 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' MA547784
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

Specificity: 13R4 recognizes wild-type E. coli beta-galactosidase.

Beta Galactosidase encodes a member of the glycosyl hydrolase 35 family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme. This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates. Mutations in this gene have been associated with GM1-gangliosidosis and Morquio B syndrome. Diseases associated with GLB1 include Gm1-Gangliosidosis, Type I and Gm1-Gangliosidosis, Type Ii.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Galactosidase Chimeric
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone 13R4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene EGFR
Gene Accession No. P00533
Gene Alias acid beta-galactosidase; AW125515; betagal; Beta-Gal; Beta-galactosidase; bgal; Bge; Bgl; Bgl-e; Bgl-s; Bgl-t; Bgs; Bgt; C130097A14Rik; EBP; Elastin receptor 1; elastin receptor 1, 67kDa; ELNR1; galactosidase beta 1; galactosidase, beta 1; GLB1; Glb-1; lactase; lysosmal hydrolase; lysosomal beta-galactosidase; lysosomal hydrolase; MPS4B; ßgal; ß-gal; ß-galactosidase
Host Species Human
Immunogen Beta-galactosidase.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1956
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.