Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Galactosidase Fusion Protein Polyclonal Antibody, eBioscience™
GREENER_CHOICE

Catalog No. 5013543
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-135-43 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-135-43 Supplier Invitrogen™ Supplier No. 14677381
Only null left
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

Description: The rabbit polyclonal antibody reacts with bBeta galactosidase. Beta-galactosidase is a large 464 kDa tetramer when assayed by western blot under nonreducing conditions and 116 kDa monomer under reducing conditions. The beta-galactosidase activity allows for the quick determination of cells expressing the lacZ gene. The lacZ gene is used as a reporter construct for determination of transfection efficiency as well as histochemical localization following transfection of eukaryotic cells. The lacZ gene product beta-galactosidase catalyzes the hydrolysis of the substrate 5-bromo-4-chloro-3-indolyl-beta-D-galactopyranoside (X-gal) to produce a blue color that is easily visualized with a microscope. Applications Reported: Purified Anti-beta galactosidase fusion proteins poly has been reported for use in immunoblotting (WB), and immunohistochemical staining. Applications Tested: Purified Anti-beta galactosidase fusion proteins poly has been tested by immunoblotting (WB). (1:400 starting dilution). It is recommended that this antibody be titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Beta Galactosidase encodes a member of the glycosyl hydrolase 35 family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme. This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates. Mutations in this gene have been associated with GM1-gangliosidosis and Morquio B syndrome. Diseases associated with GLB1 include Gm1-Gangliosidosis, Type I and Gm1-Gangliosidosis, Type Ii.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Galactosidase Fusion Protein
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with BSA, 0.02% gelatin and 0.1% sodium azide
Gene Alias acid beta-galactosidase; AW125515; betagal; Beta-Gal; Beta-galactosidase; bgal; Bge; Bgl; Bgl-e; Bgl-s; Bgl-t; Bgs; Bgt; C130097A14Rik; EBP; Elastin receptor 1; elastin receptor 1, 67kDa; ELNR1; galactosidase beta 1; galactosidase, beta 1; GLB1; Glb-1; lactase; lysosmal hydrolase; lysosomal beta-galactosidase; lysosomal hydrolase; MPS4B; ßgal; ß-gal; ß-galactosidase
Host Species Rabbit
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.