Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta Galactosidase Monoclonal Antibody (4F4F4), CoraLite488, Proteintech
SDP

Catalog No. 502334138 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-4138 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-4138 Supplier Proteintech Group Inc Supplier No. CL48866586100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Beta Galactosidase Monoclonal antibody specifically detects Beta Galactosidase in Human, Mouse, Rat samples. It is validated for Immunocytochemistry

Specifications

Antigen Beta Galactosidase
Applications Immunocytochemistry
Classification Monoclonal
Clone 4F4F4
Concentration 1 mg/mL
Conjugate CoraLite 488
Formulation PBS with 0.5% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Accession No. P16278
Gene Alias AW125515; Bge; Bgl; Bgl-e; Bgl-s; Bgl-t; Bgs; Bgt; C130097A14Rik; EBP; ELNR1; Glb-1; GLB1; MPS4B
Host Species Mouse
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2720
Target Species Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.