Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Beta-glucosidase 2 (GBA2), Rat, Unlabeled, Clone: 4A12,

Catalog No. MABN757 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN757 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN757 Supplier MilliporeSigma™ Supplier No. MABN757
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Non-lysosomal glucosylceramidase (EC 3.2.1.45; UniProt Q69ZF3; also known as Beta-glucocerebrosidase 2, Beta-glucosidase 2, Glucosylceramidase 2, NLGase) is encoded by the Gba2 (also known as Kiaa1605) gene (Gene ID 230101) in murine species. Glucosidases GBA1, GBA2, and GBA3 are structurally unrelated proteins with similar hydrolase activity that mediates glucosylceramide (GlcCer) degradation. GBA1 mediates GlcCer degradation in the lysosome lumen following its transport via the endosomal pathway. GBA2 is localized at the cytosolic surface of the ER and Golgi, with a predominant presence at the cis-Golgi in close proximity to the site of GlcCer synthesis and sphingomyelin conversion. GlcCer is also delivered to the ER through FAPP2-dependent transport and then flips to ER lumenal side by low specificity phospholipid flippases. GBA2 ER localization allows it to regulate GlcCer level at the cytosolic side of the ER and limit the amount of GlcCer from entering the ER lumenal side for higher order glycosphingolipid synthesis. Impaired GBA1 activity causes accumulation of GlcCer in lysosomes of tissue macrophages, causing liver and spleen enlargement and impairment of the central nervous system in patients with Gaucher disease. GBA2 activity is found downregulated in fibroblasts from a patient with type II Gaucher disease and fibroblasts from Gba1-deficient mice, suggesting a cross-talk between GBA1 and GBA2 that could affect the phenotype of Gaucher disease. GBA3 is a cytosolic Klotho-related protein whose physiological function is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta-glucosidase 2 (GBA2)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4A12
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat IgG1 in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Gba2, Kiaa1605
Gene Accession No. Q69ZF3
Host Species Rat
Immunogen Ovalbumin-conjugated linear peptide corresponding to a sequence from the N-terminal region of mouse beta-glucosidase 2 (GBA2).
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_766280
Test Specificity Clone 4A12 detected the beta-glucosidase 2 (GBA2) target band in testis tissue sample from wild-type, but not Gba2-knock mice (Korschen, H.G., et al. (2013). J. Biol. Chem. 288(5):3381-3393).
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.