Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Glucuronidase/GUSB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169355
Description
beta-Glucuronidase/GUSB Polyclonal specifically detects beta-Glucuronidase/GUSB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
beta-Glucuronidase/GUSB | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
beta-D-glucuronidase, beta-G1, beta-glucuronidase, BG, EC 3.2.1.31, FLJ39445, glucuronidase, beta, MPS7 | |
Rabbit | |
72 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
P08236 | |
GUSB | |
Synthetic peptides corresponding to GUSB(glucuronidase, beta) The peptide sequence was selected from the C terminal of GUSB. Peptide sequence VLGNKKGIFTRQRQPKSAAFLLRERYWKIANETRYPHSVAKSQCLENSPF. | |
Protein A purified | |
RUO | |
2990 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction