Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-Glucuronidase/GUSB Rabbit anti-Human, Clone: 4U0E2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | beta-Glucuronidase/GUSB |
---|---|
Clone | 4U0E2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
beta-Glucuronidase/GUSB Monoclonal antibody specifically detects beta-Glucuronidase/GUSB in Human samples. It is validated for Western BlotSpecifications
beta-Glucuronidase/GUSB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
2990 | |
IgG | |
Affinity purified |
4U0E2 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
beta-D-glucuronidase, beta-G1, beta-glucuronidase, BG, EC 3.2.1.31, FLJ39445, glucuronidase, beta, MPS7 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human beta-Glucuronidase/GUSB (P08236). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title