Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta glucuronidase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988179
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8179 Supplier Bioss Supplier No. BS7980R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The GUSB gene encodes beta-glucuronidase (EC 3.2.1.31), a lysosomal hydrolase involved in the stepwise degradation of glucuronic acid-containing glycosaminoglycans. It is a tetrameric glycoprotein composed of identical subunits. The GUSB gene is mutated in mucopolysaccharidosis type VII (MPS7; MIM 253220).

Specifications

Antigen beta glucuronidase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GUSB
Gene Accession No. P08236
Gene Alias Ac2-223; adipose storage deficiency; AI747421; asd; beta-D-glucuronidase; Beta-G1; Beta-glucuronidase; BG; BOS_22885; g; glucuronidase beta; glucuronidase, beta; glucuronidase, beta-like; Gur; Gus; GUSB; Gus-r; Gus-s; Gus-t; Gus-u; Gut; LOC100347788; MPS7; si:ch211-160e1.7; si:ct573103.7
Gene Symbols GUSB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GUSB/beta glucuronidase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2990
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.