Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-GlyceroPhosphate, 200 mM Solution, PROMO, Thermo Scientific Chemicals

Catalog No. J62121AEPM
Click to view available options
Quantity:
100 mL

Applications

B-Glycerolphosphate (Glycerol-2-Phosphate) is used in the development of hydrogels and scaffolds that have applications in tissue engineering and cell growth and differentiation. It has also been used as an additive in isolation mediums, similarly providing available phosphate to the isolate. It is a phosphate group donor in matrix mineralization studies that acts as a protein phosphatase inhibitor. Accelerates calcification in vascular smooth muscle cells.

Solubility

Soluble in water.

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 819-83-0
Molecular Formula C3H9Na2O6P
Molecular Weight (g/mol) 218.05 g/mol
MDL Number MFCD00002135
InChI Key UQVSFONYLAZJDB-UHFFFAOYSA-N
Synonym glycerol 2-phosphate disodium salt hydrate, disodium glycerol 2-phosphate hydrate, beta-glycerophosphoric acid disodium salt, c3h7o6p.2na.h2o, sodium glycerophosphate usan, b-glycerophosphate disodium salt hydrate, sodium 1,3-dihydroxypropan-2-yl phosphate hydrate 2:1:x, beta-glycerophosphate disodium salt hydrate, ≤1.0 mol % l-alpha-isomer, beta-glycerophosphate disodium salt hydrate, powder, bioreagent, suitable for cell culture, beta-glycerophosphate disodium salt hydrate, ≤2.4 mol % alpha-isomer based on c3h7o6pna2 with x h2o
PubChem CID 131860121
IUPAC Name 1,3-dihydroxypropan-2-yl dihydrogen phosphate;sodium
SMILES C(C(CO)OP(=O)(O)O)O.[Na].[Na]

Specifications

Color Colorless
Quantity 100 mL
Solubility Information Soluble in water.
Concentration or Composition (by Analyte or Components) 200 mM
Physical Form Liquid
Chemical Name or Material beta-Glycerophosphate

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.