Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vidan™ Diagnostics Beta-hydroxybutyrate (BHB) 21FS Reagent Kit

Catalog No. 23300109
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 18 mL R1, 2 x 5 mL R2, 1 x 3 mL Std.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-300-109 2 x 18 mL R1, 2 x 5 mL R2, 1 x 3 mL Std.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-300-109 Supplier Vidan™ Diagnostics Supplier No. 137119943018
This item is not returnable. View return policy

Beta-hydroxybutyrate (BHB) 21FS Liquid Stable, Ready-to-use Reagent

  • Prefilled Reagent Kits, makes it easy to load directly onto most clinical chemistry systems
  • Up to 12 Weeks Stability, reducing the frequency of calibrations and allows reagent to stay onboard longer
  • Bi-Level and Tri-Level Controls, color coded for your convenience (low-yellow, medium-orange, high-red)
  • Less Interference with Lipid Clearing System (LCS) that overcomes the burden of lipemia by eliminating turbidity in the sample before measurement is performed
  • The Best Indicator of DKA, βHB/Acetoacetate ratio is typically between 3:1 and 7:1 in severe ketotic states
  • FDA  Cleared
  • Expiration: 24 months from date of manufacturing

Specifications

Content And Storage Refrigerate
CE Marker Yes
Detectable Analytes beta-hydroxybutyrate
Sample Type serum and plasma
Type Quantitative Determination of Beta-Hydroxybutyrate in Serum and Plasma
Formulation Beta-Hydroxybutyrate Dehydrogenase/UV method
For Use With (Application) Open System Clinical Chemistry Analyzers
Includes BHB 21 FS Reagents (R1: 2 x 18mL; R2: 2 x 5mL; Std. 1 x 3mL)
Packaging Type (1) BHB Reagent kit
Quantity 2 x 18 mL R1, 2 x 5 mL R2, 1 x 3 mL Std.
Wavelength 340 nm
Form Liquid Ready-to-Use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.