Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Beta-hydroxybutyrate (BHB) Liquicolor™ Reagent Kit
SDP

Catalog No. 23001176 Shop All Fisherbrand Products
Click to view available options
:
BHB Reagent Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

The Fisherbrand Beta-hydroxybutyrate (BHB) Liquicolor™ test is a ready-to-use reagent that specifically measures the predominant and most acidic ketone produced during ketoacidosis. This liquid stable reagent can be used on any open channel analyzer and is more accurate and specific than ketone tablets.

  • Liquid, Ready-to-Use Reagents: Convenient liquid reagents that are pre-mixed and ready for immediate use on open channel chemistry analyzers, saving time and effort in preparation.
  • Enzymatic Detection Method: Utilizes an enzymatic detection method to ensure accuracy in detecting beta-hydroxybutyrate (BHB), providing reliable quantitative results.
  • FDA-Cleared
  • Best Indicator of Diabetic Ketoacidosis (DKA): BHB serves as the best indicator of DKA, offering valuable insights into the patient's metabolic state.
  • BHB test is available on multiple chemistry analyzer platforms such as Roche Cobas, Abbott Alinity, and QuidelOrtho Vitros with downloadable applications

Specifications

Certifications/Compliance MDSAP, ISO13485:2016, FDA-Cleared
Content And Storage Refrigerated
CE Marker No
Clia Complexity Moderate
Detectable Analytes beta-hydroxybutyrate
Sample Type Serum or Plasma
Sensitivity 99.90%
Type Beta-hydroxybutyrate (BHB) Reagent Kit
Disposable Reusable
For Use With (Application) Quantitative Measurement of Beta-Hydroxybutyrate in Serum and Plasma
For Use With (Equipment) Any open channel analyzer
Includes R1(Enzyme): 1 x 50 mL, R2 (Catalyst): 1 x 8.5 mL, STD (1 mmol/L): 1 x 3 mL
Quantity 1 Reagent Kit
Wavelength 505 nm
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.