Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vidan™ Diagnostics Beta-hydroxybutyrate (BHB) Bi-level Controls

Catalog No. 23300111
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-300-111 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-300-111 Supplier Vidan™ Diagnostics Supplier No. 595409943084
This item is not returnable. View return policy

Beta-hydroxybutyrate (BHB) Bi-level Controls can be used to perform QC of the Beta-hydroxybutyrate 21FS Reagent Kit from Vidan Diagnostics as well as other clinical chemistry BHB reagents.

  • Bi-level Control Solution for Beta-Hydroxybutyrate (BHB), (6 x 4ml)
  • Keep refrigerated
  • 60 day open vial stability
  • Perform QC as recommended by your institution
  • FDA Cleared

Specifications

Certifications/Compliance FDA cleared
Content And Storage Refrigerated
Concentration Level 1 and Level 3 controls
Detectable Analytes beta-hydroxybutyrate (BHB)
For Use With (Equipment) Open System Clinical Chemistry Analyzers
Type BHB Bi-level Controls
Includes (1) bottle level 1 control and (1) bottle of level 3 control
Level 1, 3
Quantity 1 Kit
Source Water Based with BSA
Stability 60 days once opened
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.