Learn More
Fisherbrand™ Beta-hydroxybutyrate (BHB) Liquicolor™ Reagent Kit for Beckman Synchron™ Systems (CX/LX/DX)
Shop All Fisherbrand Products

Description
- Liquid, Ready-to-Use Reagents: Convenient liquid reagents that are pre-mixed and ready for immediate use on open channel chemistry analyzers, saving time and effort in preparation.
- Enzymatic Detection Method: Utilizes an enzymatic detection method to ensure accuracy in detecting beta-hydroxybutyrate (BHB), providing reliable quantitative results.
- FDA-Cleared
- Prefilled Reagent Kits: Prefilled kits simplify the testing process by providing all necessary reagents in pre-filled Beckman Synchron™ CX/LX/DX containers, making it easy to load directly onto most clinical chemistry systems.
- Best Indicator of Diabetic Ketoacidosis (DKA): BHB serves as the best indicator of DKA, offering valuable insights into the patient's metabolic state.
- 190 Tests per Kit: Each kit provides approximately 190 tests, ensuring an ample supply for routine testing needs.
Specifications
Specifications
| Certifications/Compliance | MDSAP, ISO13485:2016, FDA-Cleared |
| Content And Storage | Refrigerated |
| CE Marker | No |
| Clia Complexity | Moderate |
| Detectable Analytes | beta-hydroxybutyrate |
| Sample Type | Serum or Plasma |
| Sensitivity | 99.90% |
| Type | Reagent Kit |
| Disposable | Reusable |
| For Use With (Application) | Quantitative Measurement of Beta-Hydroxybutyrate in Serum and Plasma |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.