Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta I Tubulin Antibody (SR1866), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32186350UL
Description
beta I Tubulin Monoclonal antibody specifically detects beta I Tubulin in Human, Mouse, Rat, Bovine, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
beta I Tubulin | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Canine, Monkey | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR1866 | |
Western Blot 1:3000-1:10000, Flow Cytometry 1:100, Immunohistochemistry 1:100-1:200, Immunocytochemistry/Immunofluorescence 1:100-1:200 | |
beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1 | |
A synthesized peptide derived from human beta I Tubulin (Uniprot #: Q9H4B7) | |
50 μg | |
Cell Biology, Cellular Markers | |
81027 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction