Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta II Tubulin B Antibody (5B2), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174037B
Description
beta II Tubulin B Monoclonal specifically detects beta II Tubulin B in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
beta II Tubulin B | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
TUBB2B | |
Recombinant human beta-Tubulin (1-445aa) purified from E. coli | |
100 μL | |
Signal Transduction | |
347733 | |
Human, Mouse | |
Purified |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
5B2 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
bA506K6.1, class II beta-tubulin isotype, DKFZp566F223, FLJ98847, MGC8685, tubulin beta-2B chain, tubulin, beta 2B, tubulin, beta polypeptide paralog | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human beta-Tubulin mAb, clone 5B2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human beta-Tubulin protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction