Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-III Tubulin Antibody (CL5814), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP26143125UL
Description
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
beta-III Tubulin | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
TUBB3 | |
This antibody was developed against a synthetic peptide corresponding to amino acids: YEDDEEESEAQGPK | |
25 μL | |
Cell Cycle and Replication, Cellular Markers, Cytoskeleton Markers, Neuroscience | |
10381 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CL5814 | |
Western Blot 1 μg/mL, Immunohistochemistry 1:50000 - 1:100000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:50000 - 1:100000 | |
beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Specificity of human beta-III Tubulin antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction