Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-III Tubulin Antibody (GT1338), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313505
This item is not returnable.
View return policy
Description
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| beta-III Tubulin | |
| Monoclonal | |
| Unconjugated | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human beta-III Tubulin. The exact sequence is proprietary. | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience | |
| 10381 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| GT1338 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction