Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-III Tubulin Antibody (TU-20), FITC, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications
Supplier: Novus Biologicals NB500620
Description
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
| beta-III Tubulin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 10 ug/ml, Flow (Intracellular) 1-3 ug/ml | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody TU-20 recognizes C-terminal peptide sequence ESESQGPK (aa 441-448) of neuron-specific human betaIII-tubulin. | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| TU-20 | |
| FITC | |
| Q13509 | |
| TUBB3 | |
| Peptide (C) 441-448 coupled to maleimide-activated keyhole limpet hemocyanin via cysteine added to the N-terminus of the neuron-specific peptide. | |
| 0.1 mg | |
| Cellular Markers | |
| 10381 | |
| Human, Mouse, Rat, All species | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction