Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-III Tubulin Mouse anti-Human, Clone: TUBB3/3732, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30738620UG
Description
beta-III Tubulin Monoclonal specifically detects beta-III Tubulin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
beta-III Tubulin | |
Monoclonal | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
A synthetic peptide (aa 437-450) of human beta-III Tubulin protein (TUBB3); coupled to KLH. | |
20 μg | |
Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience, Phospho Specific | |
10381 | |
Store at 4°C. | |
IgG2a κ |
Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
TUBB3/3732 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction