Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Beta III Tubulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585994
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585994 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585994 Supplier Invitrogen™ Supplier No. PA585994
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Beta III Tubulin Polyclonal Antibody for ICC/IF, Western Blot

The betaIII-tubulin isoform is present dominantly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta III Tubulin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene TUBB3
Gene Accession No. Q13509, Q4QRB4, Q9ERD7
Gene Alias 3200002H15Rik; beta 3 tubulin; beta tubulin 3; beta-4; betaIII-tubulin; CDCBM; CDCBM1; CFEOM3; CFEOM3A; class III beta-tubulin; FEOM3; I79_000194; LOC100341581; M(beta)3; M(beta)6; mc1R; melanocortin receptor 1; Neuron specific beta III Tubulin; neuron-specific class III beta-tubulin; TUBB; Tubb3; TUBB4; tubulin beta; tubulin beta 3 class III; Tubulin beta-3; tubulin beta-3 chain; Tubulin beta-4; Tubulin beta-4 chain; tubulin beta-III; tubulin, beta 3 class III; tubulin, beta, 3; tubulin, beta, 4; Unknown (protein for MGC:134115)
Gene Symbols TUBB3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to Human Beta III Tubulin aa 401-450.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10381, 22152, 246118
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.