Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-IV spectrin Mouse anti-Human, Rat, Clone: N393/76, MilliporeSigma™

Catalog No. MABN1727MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1727MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1727MI Supplier MilliporeSigma Supplier No. MABN1727
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Beta-IV spectrin Antibody, clone N393/76 is a highly specific mouse monoclonal antibody that targets beta-IV spectrin and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta-IV spectrin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone N393/76
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols SPTBN4,KIAA1642,SPTBN3
Host Species Mouse
Immunogen A recombinant fusion fragment corresponding to 192 amino acids from the C-terminal half of human beta-IV spectrin.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_066022
Test Specificity Clone N393/76 specifically detects human and rat beta-IV Spectrin by immunohistochemistry. it targets a sequence within 192 amino acids in the C-terminal half of human beta-IV Spectrin.
Target Species Human, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.