Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta L Crystallin Polyclonal Antibody

Catalog No. PIPA5122209
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122209 20 μL
PIPA5119054 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122209 Supplier Invitrogen™ Supplier No. PA5122209
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

beta L Crystallin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The uronate cycle functions as an alternative glucose metabolic pathway, accounting for about 5% of daily glucose catabolism. The product of this gene catalyzes the dehydrogenation of L-gulonate into dehydro-L-gulonate in the uronate cycle. The enzyme requires NAD(H) as a coenzyme, and is inhibited by inorganic phosphate. A similar gene in the rabbit is thought to serve a structural role in the lens of the eye.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta L Crystallin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CRYL1
Gene Accession No. Q99KP3, Q9Y2S2
Gene Alias 1110025H08Rik; A230106J09Rik; C85932; CRY; Cryl1; crystallin lambda 1; crystallin, lambda 1; crystallin, lamda 1; epididymis luminal protein 30; GDH; gul3dh; HEL30; lambda-CRY; Lambda-crystallin homolog; L-gulonate 3-dehydrogenase; testicular tissue protein Li 44
Gene Symbols CRYL1
Host Species Rabbit
Immunogen Recombinant protein Crystallin Lambda 1. The antigen corresponds to amino acid range 24-232 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51084, 68631
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.