Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta L Crystallin Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Catalog No. PIPA5122207
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122207 100 μL
PIPA5122208 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122207 Supplier Invitrogen™ Supplier No. PA5122207
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The uronate cycle functions as an alternative glucose metabolic pathway, accounting for about 5% of daily glucose catabolism. The product of this gene catalyzes the dehydrogenation of L-gulonate into dehydro-L-gulonate in the uronate cycle. The enzyme requires NAD(H) as a coenzyme, and is inhibited by inorganic phosphate. A similar gene in the rabbit is thought to serve a structural role in the lens of the eye.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta L Crystallin
Applications Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CRYL1
Gene Accession No. Q9Y2S2
Gene Alias 1110025H08Rik; A230106J09Rik; C85932; CRY; Cryl1; crystallin lambda 1; crystallin, lambda 1; crystallin, lamda 1; epididymis luminal protein 30; GDH; gul3dh; HEL30; lambda-CRY; Lambda-crystallin homolog; L-gulonate 3-dehydrogenase; testicular tissue protein Li 44
Gene Symbols CRYL1
Host Species Rabbit
Immunogen Met1-Gln319 (Accession Q9Y2S2), with N-terminal His Tag.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51084
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.