Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Lactamase Monoclonal Antibody (3E11.G3), Invitrogen™

Catalog No. PIMA546174
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546174 100 μg
01-671-943 500 μg
PIMA546121 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546174 Supplier Invitrogen™ Supplier No. MA546174
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10713 detects beta Lactamase from Enterobacter cloacae samples. MA1-10713 has been successfully used in ELISA applications. The MA1-10713 immunogen is Type IV beta-lactamase from Enterobacter cloacae .

The beta lactam antibiotics (penicillins and cephalosporins) are the most frequently used antimicrobial agents. All of the beta lactams are structurally related through the presence of a core beta lactam ring. Bacterial resistance to beta lactams continues to increase, primarily due to the production of beta lactamases. Beta lactamases catalyze the hydrolysis of the beta lactam bond, which destroys antibacterial activity. Bacteria that produce TEM type or SHV type beta lactamases have point mutations in structural genes that have extended the substrate specificity of these beta lactamases. As a result, many of the beta lactamase producing Gram negative pathogens have become multidrug resistant.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Lactamase
Applications ELISA
Classification Monoclonal
Clone 3E11.G3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias blaCTX-M-14; FLJ14902; G24; lactamase beta; LACTB; mitochondrial 39S ribosomal protein L56; MRPL56; Serine beta-lactamase-like protein LACTB, mitochondrial; UNQ843/PRO1781
Host Species Mouse
Immunogen Type IV beta-lactamase from Enterobacter cloacae.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.