Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Beta-Lactamase (TEM-1) Recombinant Protein

Catalog No. PV3575
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3575 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV3575 Supplier Invitrogen™ Supplier No. PV3575
Add to Cart
Edge
Add to Cart

Recombinant Protein

TEM-1 beta-lactamase is the most widespread enzyme to confer beta-lactam antibiotic resistance to gram-negative bacteria. It is a monomeric protein that requires no metals or cofactors for activity. TEM-1 beta-lactamase efficiently hydrolyzes most penicillin and cephalosporin substrates, including nitrocefin and fluorescent derivates. This protein is intended to be used as a positive control for enzyme-dependent substrate hydrolysis and for use on immunoblots. Other uses include kinetic studies with novel substrates or identification of inhibitors of beta-lactamase as part of an antimicrobial program. 1165 micromoles of penicillin G hydrolyzed per minute per milligram of total protein.

CD248 (CD248 Molecule) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include calcium ion binding and extracellular matrix binding. An important paralog of this gene is CD93. CD248 may play a role in tumor angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9HCU0
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 200mM potassium phosphate with 50% glycerol and no preservative; pH 7
Gene ID (Entrez) 57124
Molecular Weight (g/mol) 38 kDa
Name Beta-Lactamase (TEM-1)
pH Range 7
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C
Regulatory Status RUO
Gene Alias 2610111G01Rik; AI842296; CD164 sialomucin-like 1; CD164L1; Cd248; CD248 antigen, endosialin; CD248 molecule; CD248 molecule, endosialin; endosialin; MGC119478; MGC119479; TEM1; tumor endothelial marker 1
Common Name CD248
Gene Symbol CD248
Biological Activity 1165 micromoles of penicillin G hydrolyzed / minute / milligram of total protein.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Content And Storage -80°C
Activity 1165 micromoles of penicillin G hydrolyzed per minute per milligram of total protein.
Shipping Condition Dry Ice
Expression System E. coli
Form Liquid
Protein Subtype Other Proteins
Research Category Drug Discovery
Product Line GeneBLAzer
Purity or Quality Grade 95% as determined by a Coomassie ™ blue stained SDS-PAGE gel. Calculated from the protein sequence(s).
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.