Learn More
beta Lactoglobulin Rabbit anti-Bovine, Polyclonal, Nordic

Rabbit Polyclonal Antibody
Supplier: Vector Laboratories RAB/BLGL

Description
Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2). Repeated thawing and freezing should be avoided. If a slight precipitation occurs upon storage, this should be removed by centrifugation. It will not affect the performance of the antiserum. Diluted antiserum should be stored at 4 C, not refrozen, and preferably used the same day. In immunoelectrophoresis use 2 ul guinea pig plasma or equivalent against 120 ul antiserum. In double radial immunodiffusion use a rosette arrangement with 10 ul antiserum in 3 mm diameter center well and 2 ul plasma samples (neat and serially diluted) in 2 mm diameter peripheral wells. In single radial immunodiffusion use 1 percent antiserum in the gel.
beta Lactoglobulin Polyclonal antibody specifically detects beta Lactoglobulin in Bovine samples. It is validated for ImmunoelectrophoresisSpecifications
beta Lactoglobulin | |
Polyclonal | |
whole serum with no preservative; pH | |
P02754 | |
Rabbit | |
1 mL | |
Primary | |
Bovine | |
Antibody | |
IgG |
Immunoelectrophoresis | |
Unconjugated | |
PAEP | |
beta lactoglobulin C; beta lactoglobulin D; Beta-lactoglobulin; beta-LG; BLG; bLG C; Bos d 5; lactoglobulin, beta; LGB; PAEP; progestagen-associated endometrial protein | |
Bovine beta lactoglobulin | |
RUO | |
280838 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.