Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-NGF Antibody (016), Alexa Fluor™ 647

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290552AF647
Description
beta-NGF Monoclonal specifically detects beta-NGF in Mouse samples. It is validated for ELISA.Specifications
beta-NGF | |
Monoclonal | |
Alexa Fluor 647 | |
Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse beta-NGF (NP_001106168.1; Ser122-Gly241). | |
0.1 mL | |
Cancer, Neuroscience | |
4803 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
16 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction