Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-NGF Mouse, HRP, Clone: 25623, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12010513H
Description
beta-NGF Monoclonal specifically detects beta-NGF in Human, Mouse, Rat samples. It is validated for ELISA.Specifications
beta-NGF | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NGF | |
Recombinant full length protein (Human). | |
0.1 mL | |
Cancer, Neuroscience | |
4803 | |
Human, Mouse, Rat | |
Purified |
ELISA | |
25623 | |
ELISA | |
Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NGF beta (25623.1) | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction