Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Nicotinamide adenine dinucleotide phosphate disodium salt, 97%

Catalog No. p-7042445
Click to view available options
Quantity:
250 mg
1 g

CAS: 24292-60-2 | C21H26N7Na2O17P3 | 787.37 g/mol

Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system.

Solubility
Soluble in water (50 mg/ml), 0.001% aqueous buffer, pH 7.0, methanol, and ethanol (less soluble). Insoluble in ether, and ethyl acetate.

Notes
Protect from light and moisture. Keep cool and dry. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24292-60-2
Molecular Formula C21H26N7Na2O17P3
Molecular Weight (g/mol) 787.37
MDL Number MFCD00065390
InChI Key WSDDJLMGYRLUKR-MWLGWILCNA-L
Synonym nadp disodium salt, beta-nicotinamide adenine dinucleotide phosphate disodium salt
PubChem CID 131851653
IUPAC Name [[(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-3-hydroxy-4-phosphonooxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
SMILES [Na+].[Na+].NC(=O)C1=CC=C[N+](=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP(O)([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O

Specifications

Color White
Quantity 250 mg
Solubility Information Soluble in water (50mg/ml),0.001% aqueous buffer,pH 7.0,methanol,and ethanol (less soluble). Insoluble in ether,and ethyl acetate.
Formula Weight 787.37
Percent Purity 97%
Physical Form Powder
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate disodium salt
EINECSNumber 246-129-8
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.