Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-PhosphoglucomutaseGoat anti-Bacteria, Polyclonal, Rockland™
SDP

Catalog No. 501989988
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9988 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9988 Supplier Rockland Immunochemicals Supplier No. 100101237
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-goat serum, purified and partially purified b-Phosphoglucomutase [Lactococcus lacti]. Cross reactivity against b -Phosphoglucomutase from other sources is unknown.

Beta-Phosphoglucomutase catalyzes the interconversion of D-glucose 1-phosphate (G1P) and D-glucose 6-phosphate (G6P), forming beta-D-glucose 1,6-(bis)phosphate (beta-G16P) as an intermediate. The beta-phosphoglucomutase (Beta-PGM) acts on the beta-C1 anomer of G1P. Glucose or lactose are used in preference to maltose, which is only utilized after glucose or lactose has been exhausted. It plays a key role in the regulation of the flow of carbohydrate intermediates in glycolysis and the formation of the sugar nucleotide UDP-glucose. The catalysis proceeds via a phosphoenzyme formed by reaction of an active-site nucleophile with the cofactor glucose 1,6-diphosphate (G1,6-diP). The phosphorylated mutase binds either G1P or G6P and transfers the phosphoryl group to the C6OH or C1OH, respectively.

Specifications

Antigen beta-Phosphoglucomutase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 90 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Goat
Immunogen Beta Phosphoglucomutase [Lactococcus lacti]
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.