Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta-TrCP1/BTRC Antibody (OTI2H2), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271402JF549
Description
beta-TrCP1/BTRC Monoclonal specifically detects beta-TrCP1/BTRC in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
beta-TrCP1/BTRC | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BTRC | |
Recombinant protein fragment corresponding to amino acids 52-354 of human beta-TrCP1/BTRC (NP_378663) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI2H2 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
beta-transducin repeat containing, betaTrCP, BETA-TRCP, beta-TrCP1, bTrCP, bTrCP1, E3RSIkappaB, F-box and WD repeats protein beta-TrCP, F-box/WD repeat-containing protein 1A, FBW1A, FBXW1, FBXW1ABTRCP, Fwd1, MGC4643, pIkappaBalpha-E3 receptor subunit | |
Mouse | |
Affinity Purified | |
Signal Transduction, Vision | |
8945.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction