Learn More
beta TrCP2Rabbit anti-Bovine, Canine, Chicken, Human, Monkey, Mouse, Polyclonal, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 600401483
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody reacts with human betaTrCP2 protein. A BLAST analysis was used to suggest cross-reactivity with betaTrCP2 from human, mouse, dog, bovine, chimpanzee, chicken and horse based on a 100% homology with the immunizing sequence. Cross-reactivity with betaTrCP2 from other sources has not been determined.
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbws class and, in addition to an F-box, contains multiple WD40 repeats. This gene contains at least 14 exons, and its alternative splicing generates 3 transcript variants diverging at the presence/absence of two alternate exons.Specifications
| beta TrCP2 | |
| Polyclonal | |
| Unconjugated | |
| Fbxw11 | |
| 2310065A07Rik; AA536858; beta-transducin repeat-containing protein 2; BTRC2; BTRCP2; F-box and WD repeat domain containing 11; F-box and WD repeats protein beta-TrCP2; F-box and WD-40 domain protein 11; F-box and WD-40 domain protein 1B; F-box protein Fbw1b; F-box/WD repeat-containing protein 11; F-box/WD repeat-containing protein 1B; Fbw11; Fbw1b; FBXW11; Fbxw1b; Homologous to Slimb protein; HOS; KIAA0696 | |
| Rabbit | |
| RUO | |
| 103583, 23291, 416181, 479291, 540642 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.8 mg/mL | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| Q5SRY7, Q9UKB1 | |
| Fbxw11 | |
| 100 μg | |
| Primary | |
| Bovine, Canine, Chicken, Human, Monkey, Mouse | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.