Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ beta Tubulin 2,4 Recombinant Rabbit Monoclonal Antibody (ST52-04)
GREENER_CHOICE

Catalog No. PIMA532163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532163 Supplier Invitrogen™ Supplier No. MA532163
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Tubulin 2,4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone ST52-04
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene TUBB4A
Gene Accession No. P04350, P68371, P68372, P85108, Q13885, Q3KRE8, Q6P9T8, Q7TMM9, Q9BVA1, Q9CWF2, Q9D6F9
Gene Alias 2410129E14Rik; 4930542G03Rik; AI325297; bA506K6.1; Beta2; beta-5; beta-tubulin T beta15 (aa 1-445); brdp; CDCBM5; class II beta-tubulin isotype; class IIa beta-tubulin; class IIb beta-tubulin; class IVb beta tubulin; class IVb beta-tubulin; dystonia 4, torsion (autosomal dominant); DYT4; M(beta)2; M(beta)4; novel tubulin protein; PMGYSA; RGD1309427; t beta-15; TUBB; Tubb2; TUBB2A; tubb2a {ECO:0000250; Tubb2b; TUBB2C; Tubb2c1; Tubb4; Tubb4a; Tubb4b; TUBB5; Tubulin 5 beta; tubulin beta; tubulin beta 2A class IIa; tubulin beta 2B class IIb; tubulin beta 4A class IVa; tubulin beta 4B class IVb; Tubulin beta class IIa; Tubulin beta-2 chain; tubulin beta-2A chain; Tubulin beta-2B chain; tubulin beta-2C chain; Tubulin beta-4 chain; tubulin beta-4A chain; Tubulin beta-4B chain; tubulin, beta 2; tubulin, beta 2A class IIa; tubulin, beta 2B class IIb; tubulin, beta 2C; tubulin, beta 2c1; tubulin, beta 4; tubulin, beta 4 class IVa; tubulin, beta 4A class IVa; tubulin, beta 4B class IVb; tubulin, beta polypeptide; tubulin, beta polypeptide 2; tubulin, beta polypeptide paralog; tubulin, beta, 2; tubulin, beta, 5; UniProtKB:Q7TMM9}
Gene Symbols Tubb2a, Tubb2b, TUBB4A, TUBB4B
Host Species Rabbit
Immunogen Synthetic peptide within Human beta II Tubulin aa 407-445.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10382, 10383, 22151, 22153, 227613, 291081, 29213, 296554, 347733, 498736, 7280, 73710
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.