Learn More
Invitrogen™ beta Tubulin 2 Recombinant Rabbit Monoclonal Antibody (1R3R9)

Description
beta Tubulin 2 Recombinant Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | beta Tubulin 2 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 1R3R9 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TUBB2A |
| Gene Accession No. | P85108, Q13885, Q3KRE8, Q7TMM9, Q9BVA1, Q9CWF2 |
| Gene Alias | 2410129E14Rik; 2t; b2 tubulin; B2t; bA506K6.1; beta 2 tubulin; BETA 85D; beta tubulin; beta(2)Tu; beta(2)Tub; beta(2)Tubulin; beta;Tub85D; beta[[2]]-tubulin; BETA2; beta2 tubulin; beta2t; beta2tub; beta2-tub; beta2tubulin; Beta-2-tubulin; beta2-tubulin; beta85D; betaTub; beta-tub; betaTub2; BetaTub85D; beta-tub85D; betaTub85D-PA; beta-Tubulin; beta-Tubulin 85D; beta-Tubulin at 85D; beta-tubulin T beta15 (aa 1-445); betaTubulin85D; beta-tubulin85D; brdp; CDCBM5; CG9359; CG9359-PA; class II beta-tubulin isotype; class IIa beta-tubulin; class IIb beta-tubulin; D.m.BETA-85D; dJ40E16.7; Dmbeta2; Dmel\CG9359; Dmel_CG9359; DTB4; GH02051p; M(beta)2; ms(3)KK[D]; PMGYSA; RGD1309427; t beta-15; testis-specific beta-tubulin; Tub; Tub2A; TUBB; TUBB2; TUBB2A; tubb2a {ECO:0000250; Tubb2b; TubB85D; tubulin; tubulin beta; tubulin beta 2A class IIa; tubulin beta 2B class IIb; Tubulin beta class IIa; Tubulin beta-2 chain; Tubulin beta-2A chain; tubulin beta-2B chain; tubulin, beta 2; tubulin, beta 2A class IIa; tubulin, beta 2B class IIb; tubulin, beta polypeptide; tubulin, beta polypeptide 2; tubulin, beta polypeptide paralog; UniProtKB:Q7TMM9} |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.