Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Tubulin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107300 Supplier Thermo Scientific Supplier No. PA5107300
Add to Cart
Add to Cart

beta Tubulin 2 Polyclonal Antibody

beta Tubulin 2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Tubulin 2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene TUBB2A
Gene Accession No. Q13885, Q9BVA1
Gene Alias 2410129E14Rik; 2t; b2 tubulin; B2t; bA506K6.1; beta 2 tubulin; BETA 85D; beta tubulin; beta(2)Tu; beta(2)Tub; beta(2)Tubulin; beta;Tub85D; beta[[2]]-tubulin; BETA2; beta2 tubulin; beta2t; beta2tub; beta2-tub; beta2tubulin; Beta-2-tubulin; beta2-tubulin; beta85D; betaTub; beta-tub; betaTub2; BetaTub85D; beta-tub85D; betaTub85D-PA; beta-Tubulin; beta-Tubulin 85D; beta-Tubulin at 85D; beta-tubulin T beta15 (aa 1-445); betaTubulin85D; beta-tubulin85D; brdp; CDCBM5; CG9359; CG9359-PA; class II beta-tubulin isotype; class IIa beta-tubulin; class IIb beta-tubulin; D.m.BETA-85D; dJ40E16.7; Dmbeta2; Dmel\CG9359; Dmel_CG9359; DTB4; GH02051p; M(beta)2; ms(3)KK[D]; PMGYSA; RGD1309427; t beta-15; testis-specific beta-tubulin; Tub; Tub2A; TUBB; TUBB2; TUBB2A; tubb2a {ECO:0000250; Tubb2b; TubB85D; tubulin; tubulin beta; tubulin beta 2A class IIa; tubulin beta 2B class IIb; Tubulin beta class IIa; Tubulin beta-2 chain; Tubulin beta-2A chain; tubulin beta-2B chain; tubulin, beta 2; tubulin, beta 2A class IIa; tubulin, beta 2B class IIb; tubulin, beta polypeptide; tubulin, beta polypeptide 2; tubulin, beta polypeptide paralog; UniProtKB:Q7TMM9}
Gene Symbols TUBB2A, TUBB2B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Human TUBB2B.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347733, 7280
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.