Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Tubulin Antibody (1B12), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250061MFV500
Description
beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Single Cell WesternSpecifications
beta Tubulin | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence, Single Cell Western | |
1B12 | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Single Cell Western | |
beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
Pig brain beta Tubulin preparation. [UniProt# Q767L7] | |
0.1 mL | |
Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
203068 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction