Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Tubulin Antibody (4E4), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250047IR
Description
beta Tubulin Monoclonal specifically detects beta Tubulin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
beta Tubulin | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TUBB | |
Pig brain beta Tubulin. [UniProt# Q767L7] | |
0.1 mL | |
Apoptosis, Breast Cancer, Cellular Markers, Cytoskeleton Markers, Signal Transduction | |
203068 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
40000 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction