Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Tubulin Antibody (BT7R) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP2378300.025MG
Description
beta Tubulin Monoclonal specifically detects beta Tubulin in Human, Mouse, Rat, Avian, Chicken, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), CyTOF-ready.Specifications
beta Tubulin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 - 1:5000, Flow Cytometry 1 μg/test, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000, Immunohistochemistry-Paraffin 1:10 - 1:500, Flow (Intracellular), CyTOF-ready | |
beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Detects beta-tubulin in human, monkey, mouse, rat, rabbit and chicken samples with ELISA, Western blot and immunofluorescence applications. | |
Store at -20°C. Avoid freeze/thaw cycles. | |
IgG2a |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
BT7R | |
Unconjugated | |
PBS (pH 7.2) with No Preservative | |
TUBB | |
KLH conjugated peptide of the N-terminus of Beta-Tubulin. | |
0.025 mg | |
Apoptosis, Breast Cancer, Cellular Markers, Cytoskeleton Markers, Signal Transduction | |
203068 | |
Human, Mouse, Rat, Avian, Chicken, Primate, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction