Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Tubulin Antibody (S11B), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281064H
Description
beta Tubulin Monoclonal specifically detects beta Tubulin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
beta Tubulin | |
Monoclonal | |
HRP | |
PBS | |
TUBB | |
Human MBP (microtubule-binding protein). | |
0.1 mL | |
Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
203068 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
S11B | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Binds specifically to human beta Tubulin (no crossreactivity with alpha-tubulin). | |
Store at 4°C in the dark. | |
IgG λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction