Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Tubulin Antibody (SAP.4G5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12011312
Description
beta Tubulin Monoclonal specifically detects beta Tubulin in Human, Mouse, Rat, Bovine, Canine, Chicken, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
beta I Tubulin | |
Monoclonal | |
Unconjugated | |
beta tubulin 1, class VI, dJ543J19.4, tubulin beta-1 chain, tubulin, beta 1 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
No reactivity with other tubulin isotypes is observed. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SAP.4G5 | |
Western Blot 1:20000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
TUBB1 | |
Synthetic peptide corresponding to the C-terminal sequence of beta-tubulin isotype I, coupled to BSA. | |
0.1 mL | |
Cytoskeleton Markers | |
81027 | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Xenopus species | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction