Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Tubulin III, Neuronal Mouse anti-Rat, Human, Mouse, Bovine, Clone: 2G10, MilliporeSigma™

Mouse Monoclonal Antibody

$188.90 - $485.00

Specifications

Antigen beta-Tubulin III, Neuronal
Clone 2G10
Concentration 0.5 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05559I25UL
View Documents
MilliporeSigma
05559I25UL
25 μL
Each for $188.90
Only null left
Add to Cart
 
05559I100UL
View Documents
MilliporeSigma
05559I100UL
100 μL
Each for $485.00
Only null left
Add to Cart
 
Description

Description

beta-Tubulin III, Neuronal Monoclonal antibody specifically detects beta-Tubulin III, Neuronal in Rat, Human, Mouse, Bovine samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

beta-Tubulin III, Neuronal
0.5 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2a κ
Protein G purified
2G10
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Unconjugated
Mouse
Rat, Human, Mouse, Bovine
KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Tubulin beta-III.
Primary
Stable for 1 year at +2°C to +8°C from date of receipt.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.