Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-Tubulin IV Monoclonal Antibody (ABM871), Abbomax

Catalog No. 502204140
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4140 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4140 Supplier Abbomax Inc Supplier No. 603110
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Cerebellum, Cellular location: Cytoplasmic.

Beta-Tubulin IV Monoclonal antibody specifically detects Beta-Tubulin IV in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Beta-Tubulin IV
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone ABM871
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene TUBB4A
Gene Accession No. P04350
Gene Alias AI325297; beta-5; class IVa beta-tubulin; dystonia 4, torsion (autosomal dominant); DYT4; hypothetical protein LOC540236; M(beta)4; QtrA-11757; Tubb; TUBB4; Tubb4a; TUBB5; Tubulin 5 beta; tubulin beta; tubulin beta 4A class IVa; tubulin beta-4 chain; Tubulin beta-4A chain; tubulin, beta 4; tubulin, beta 4 class IVa; tubulin, beta 4A class IVa; tubulin, beta, 5; unnamed protein product
Host Species Mouse
Immunogen A Synthetic peptide sequence corresponding to the C-terminus of human beta-Tubulin-IV
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10382
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.