Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ 18 HPLC Columns, 5μm Particle Size

Catalog No. 71505052130 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
50 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
4.6 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
71505052130 50 mm 2.1 mm
71505154630 150 mm 4.6 mm
71505254630 250 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 71505052130 Supplier Thermo Scientific™ Supplier No. 71505052130
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Available in limited dimensions. Please contact Customer Service for more details.

Thermo ScientificTM BetaBasicTM 18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.

  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

Ordering Information:

Other column dimensions are also available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
For Use With LC/MS, APCI
Length (Metric) 50 mm
Particle Size 5 μm
pH 2 to 12
Pore Size 150 Å
Surface Area 200 m²/g
USP Type L1
Stationary Phase C18
Product Line Beta
Packing Material Silica
Carbon Load 13%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.