Learn More
Thermo Scientific™ BetaBasic™ Javelin™ C18 Reversed Phase HPLC Column, 5 μm, 4 mm x 20 mm
Shop All Thermo Scientific ProductsDescription
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
- Highly efficient and reproducible Thermo Scientific columns
- 150Å pore size for small molecules, peptides and protein digests
- Superb pH stability
Specifications
Specifications
| Column Format | Guard Column |
| Diameter (Metric) | 4 mm |
| Length (Metric) | 20 mm |
| Particle Size | 5 μm |
| Pore Size | 150 Å |
| USP Type | L1 |
| Stationary Phase | C18 |
| Product Line | Beta |
| Packing Material | High Purity, Base-Deactivated Type B Silica |
| Carbon Load | 13% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.